Learn More
EIF3E Mouse anti-Human, Unconjugated, Clone: 1C6C8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67095-1-IG-20UL
 View more versions of this product
Product Code. 16829525
  • £135.00 / 20µL
Description
The EIF3E antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF3E. This antibody recognizes human, mouse, rat antigen. The EIF3E antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
EIF3E
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, ELISA
1C6C8
Unconjugated
20 μL
3646
IgG1