missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3D Rabbit anti-Human, Mouse, Yeast, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10219-1-AP-150UL
This item is not returnable.
View return policy
Description
eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.Specifications
| EIF3D | |
| Polyclonal | |
| Unconjugated | |
| EIF3D | |
| eIF 3 zeta, eIF3 p66, eIF3 zeta, EIF3D, EIF3S7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55944, 8664 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O15371, O70194 | |
| Eif3d | |
| EIF3D Fusion Protein Ag0268 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Yeast | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction