Learn More
EIF3D Mouse anti-Human, CoraLite 594, Clone: 2C5A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66024-100UL
Description
The EIF3D antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human EIF3D. This antibody recognizes human, mouse, rat antigen. The EIF3D antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| EIF3D | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 8664 | |
| IgG2b |
| Immunofluorescence | |
| 2C5A3 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |