missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97518-50ul
This item is not returnable.
View return policy
Description
EIF3D Polyclonal antibody specifically detects EIF3D in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| EIF3D | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| eIF3d, eIF3-p66, EIF3S7eIF3 p66, eIF-3-zeta, eIF3-zeta, Eukaryotic translation initiation factor 3 subunit 7, eukaryotic translation initiation factor 3 subunit D, eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa, eukaryotic translation initiation factor 3, subunit D, MGC126526, MGC17258, translation initiation factor eIF3 p66 subunit | |
| Produced in rabbits immunized with E. coli-derived Human EIF3D fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8664 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction