missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3D Antibody (2D11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42964
This item is not returnable.
View return policy
Description
EIF3D Monoclonal antibody specifically detects EIF3D in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| EIF3D | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 8664 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 2D11 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| eIF3d, eIF3-p66, EIF3S7eIF3 p66, eIF-3-zeta, eIF3-zeta, Eukaryotic translation initiation factor 3 subunit 7, eukaryotic translation initiation factor 3 subunit D, eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa, eukaryotic translation initiation factor 3, subunit D, MGC126526, MGC17258, translation initiation factor eIF3 p66 subunit | |
| eukaryotic translation initiation factor 3, subunit D - (Uniprot# O15371) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction