missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3c Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527527
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (93%), Xenopus laevis (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
Specifications
| eIF3c | |
| Polyclonal | |
| Unconjugated | |
| Eif3c | |
| 110kDa; 3230401O13Rik; cell migration-inducing protein 17; eIF3 p110; eIF3 p110 {ECO:0000255; eIF3c; eIF3c {ECO:0000255; EIF3CL; eIF3-p110; EIF3S8; eukaryotic translation initiation factor 3 subunit 8; eukaryotic translation initiation factor 3 subunit 8 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit C; eukaryotic translation initiation factor 3 subunit C {ECO:0000255; eukaryotic translation initiation factor 3, subunit 8 (110 kDa); eukaryotic translation initiation factor 3, subunit 8 (110kD); eukaryotic translation initiation factor 3, subunit 8, 110kDa; eukaryotic translation initiation factor 3, subunit C; extra-toes spotting; HAMAP-Rule:MF_03002}; NIPIL(A3); NipilA3; NIPI-like protein; Xs; Xsl | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.9 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q99613 | |
| Eif3c | |
| Recombinant protein encompassing a sequence within the C-terminus region of human EIF3S8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction