missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3b Recombinant Rabbit Monoclonal Antibody (23GB3830), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | eIF3b |
|---|---|
| Clone | 23GB3830 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089967
![]() |
Invitrogen™
MA552588 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087700
|
Invitrogen™
MA552589 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF3b Recombinant Monoclonal antibody specifically detects eIF3b in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| eIF3b | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P55884, Q4G061, Q8JZQ9 | |
| 27979, 288516, 8662 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3830 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AL033316; AL033334; AL033369; AW208965; D5Wsu45e; eIF3 p110; eIF3 p116; EIF3B; eIF3b {ECO:0000255; eIF-3-eta; EIF3-ETA; eIF-3-eta {ECO:0000255; EIF3-P110; EIF3-P116; EIF3S9; eukaryotic translation initiation factor 3 subunit 9; eukaryotic translation initiation factor 3 subunit 9 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit B; eukaryotic translation initiation factor 3 subunit B {ECO:0000255; eukaryotic translation initiation factor 3, subunit 9 (eta); eukaryotic translation initiation factor 3, subunit 9 (eta, 116kD); eukaryotic translation initiation factor 3, subunit 9 eta, 116kDa; eukaryotic translation initiation factor 3, subunit B; HAMAP-Rule:MF_03001}; hPrt1; MGC104664; MGC131875; PRT1; Prt1 homolog | |
| A synthetic peptide of human eIF3B (151-200AA). | |
| Primary | |
| -20°C | |
| EIF3B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title