missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10319-1-AP-20UL
This item is not returnable.
View return policy
Description
The cyclin-dependent kinase inhibitor 1C (CDKN1C), also known as p57KIP2, is a tight-binding, strong inhibitor of several G1 cyclin/CDK complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndrome, a cancer-predisposing syndrome, suggesting that this gene is a tumor suppressor candidate. CDKN1C is widely expressed during organogenesis with its expression declining after maturity. It is part of the CIP/KIP family of cyclin-dependent inhibitors (CKIs) which also includes CDKN1A and CDKN1B.Specifications
| EIF3B | |
| Polyclonal | |
| Unconjugated | |
| EIF3B | |
| eIF 3 eta, EIF3 ETA, EIF3 P110, EIF3 P116, EIF3B, EIF3S9, hPrt1, PRT1, Prt1 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27979, 288516, 8662 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P55884, Q4G061, Q8JZQ9 | |
| Eif3b | |
| EIF3B Fusion Protein Ag0386 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction