missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3B Rabbit anti-Human, Mouse, Rat, Clone: 4T9V1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16765-100UL
This item is not returnable.
View return policy
Description
EIF3B Monoclonal antibody specifically detects EIF3B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| EIF3B | |
| Monoclonal | |
| Unconjugated | |
| eIF3 p110, eIF3 p116, eIF3b, eIF-3-eta, EIF3-P110, EIF3-P116, EIF3S9EIF3-ETA, Eukaryotic translation initiation factor 3 subunit 9, eukaryotic translation initiation factor 3 subunit B, eukaryotic translation initiation factor 3, subunit 9 (eta, 116kD), eukaryotic translation initiation factor 3, subunit 9 eta, 116kDa, eukaryotic translation initiation factor 3, subunit B, MGC104664, MGC131875, Prt1 homolog, PRT1hPrt1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 101-200 of human EIF3B (P55884). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| 4T9V1 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8662 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction