missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3a Recombinant Rabbit Monoclonal Antibody (23GB830), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | eIF3a |
|---|---|
| Clone | 23GB830 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089787
![]() |
Invitrogen™
MA553164 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089434
|
Invitrogen™
MA553165 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF3a Recombinant Monoclonal antibody specifically detects eIF3a in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| eIF3a | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P23116, Q14152, Q1JU68 | |
| 13669, 292148, 8661 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB830 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 150/170; A830012B05Rik; Centrosomin; centrosomin homolog; Csma; cytoplasmic protein p167; Eif3; eIF3 p167; eIF3 p180; eIF3 p185; EIF3, p180 subunit; eIF3a; eIF3a {ECO:0000255; eIF3-p170; EIF3S10; eIF-3-theta; eIF3-theta; eIF-3-theta {ECO:0000255; eukaryotic translation initiation factor 3 subunit 10; EUKARYOTIC TRANSLATION INITIATION FACTOR 3 SUBUNIT 10 (EIF-3 THETA) (EIF3 P167) (EIF3 P180) (EIF3 P185) (P162 PROTEIN) (CENTROSOMIN); eukaryotic translation initiation factor 3 subunit 10 {ECO:0000255; eukaryotic translation initiation factor 3 subunit A; eukaryotic translation initiation factor 3 subunit A {ECO:0000255; eukaryotic translation initiation factor 3, subunit 10 (theta); eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD); eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD); eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa; eukaryotic translation initiation factor 3, subunit 10, 170kD; eukaryotic translation initiation factor 3, subunit A; HAMAP-Rule:MF_03000}; KIAA0139; mKIAA0139; p162; P167; p180; p185; TIF32; ZH12 | |
| A synthetic peptide of human eIF3A (1100-1154AA). | |
| Primary | |
| -20°C | |
| EIF3A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title