missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27665-1-AP-20UL
This item is not returnable.
View return policy
Description
The German cDNA consortium.Specifications
| EIF3A | |
| Polyclonal | |
| Unconjugated | |
| EIF3A | |
| eIF 3 theta, EIF3, eIF3 p167, eIF3 p170, eIF3 p180, eIF3 p185, eIF3 theta, EIF3A, EIF3S10, KIAA0139, P167, p180, p185, TIF32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8661 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14152 | |
| EIF3A | |
| EIF3A Fusion Protein Ag24752 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction