missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3A, Mouse anti-Human, Clone: 2G4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008661-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: MPAYFQRPENALKRANEFLEVGKKQPALDVLYDVMKSKKHRTWQKIHEPIMLKYLELCVDLRKSHLAKEGLYQYKSpecifications
| EIF3A | |
| Monoclonal | |
| Unconjugated | |
| eukaryotic translation initiation factor 3, subunit A | |
| EIF3/EIF3S10/KIAA0139/P167/TIF32/eIF3-p170/eIF3-theta/p180/p185 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2G4 | |
| In 1x PBS, pH 7.4 | |
| NM_003750 | |
| EIF3A | |
| EIF3A (NP_003741, 1 a.a. ∼ 75 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8661 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction