missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
eIF2B1 Polyclonal antibody specifically detects eIF2B1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | eIF2B1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_001405.1 |
| Gene Alias | EIF2B, EIF-2B, eIF-2B GDP-GTP exchange factor subunit alpha, EIF-2Balpha, EIF2BAMGC117409, eukaryotic translation initiation factor 2B, subunit 1 (alpha, 26kD), eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa, MGC125868, MGC125869, translation initiation factor eIF-2B subunit alpha |
| Host Species | Mouse |
| Immunogen | EIF2B1 (NP_001405.1, 1 a.a. - 305 a.a.) full-length human protein. MDDKELIEYFKSQMKEDPDMASAVAAIRTLLEFLKRDKGETIQGLRANLTSAIETLCGVDSSVAVSSGGELFLRFISLASLEYSDYSKCKKIMIERGELFLRRISLSRNKIADLCHTFIKDGATILTHAYSRVVLRVLEAAVAAKKRFSVYVTESQPDLSGKKMAKALCHLNVPVTVVLDAAVGYIMEKADLVIVGAEGVVENGGIINKIGTNQMAVCAKAQNKPFYVVAESFKFVRLFPLNQQDVPDKFKYKADTLKVAQTGQDLKEEHPWVDYTAPSLITLLFTDLGVLTPSAVSDELIKLYL |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?