missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF2b epsilon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584808
This item is not returnable.
View return policy
Description
Immunogen sequence: MDDIKVFQNE VLGTLQRGKE ENISCDNLVL EINSLKYAYN ISLKEVMQVL SHVVLEFPLQ QMDSPLDSSR YCALLL Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 84%.
This gene encodes one of five subunits of eukaryotic translation initiation factor 2B, a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
Specifications
| eIF2b epsilon | |
| Polyclonal | |
| Unconjugated | |
| Eif2b5 | |
| C81315; CACH; CLE; EIF-2B; eIF-2B GDP-GTP exchange factor subunit epsilon; EIF2B5; EIF2BE; EIF2Bepsilon; eukaryotic translation initiation factor 2B subunit epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa; initiation factor eIF-2Be; LVWM; Translation initiation factor eIF-2B subunit epsilon | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8893 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13144 | |
| Eif2b5 | |
| Recombinant protein corresponding to Human EIF2B5. Recombinant protein control fragment (Product #RP-107686). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction