missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2B epsilon Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56270-25ul
This item is not returnable.
View return policy
Description
eIF2B epsilon Polyclonal specifically detects eIF2B epsilon in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| eIF2B epsilon | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| CACH, CLE, EIF-2B, eIF-2B GDP-GTP exchange factor subunit epsilon, EIF2BE, EIF2Bepsilon, eukaryotic translation initiation factor 2B, subunit 5 (epsilon, 82kD), eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa, LVWM, translation initiation factor eIF-2B subunit epsilon | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| EIF2B5 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SILEENVLLGSGTVIGSNCFITNSVIGPGCHIGDNVVLDQTYLWQGVRVAAGAQIHQSLLCDNAEVKERVTLKPRSVLTSQVVVGPNITLPEGSVISLHPPD | |
| 25 μL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 8893 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction