missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2B epsilon Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35116-20ul
This item is not returnable.
View return policy
Description
eIF2B epsilon Polyclonal antibody specifically detects eIF2B epsilon in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| eIF2B epsilon | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| CACH, CLE, EIF-2B, eIF-2B GDP-GTP exchange factor subunit epsilon, EIF2BE, EIF2Bepsilon, eukaryotic translation initiation factor 2B, subunit 5 (epsilon, 82kD), eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa, LVWM, translation initiation factor eIF-2B subunit epsilon | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 442-721 of human eIF2B epsilon (eIF2B epsilon (EIF2B5)) (NP_003898.2).,, Sequence:, TLPEGSVISLHPPDAEEDEDDGEFSDDSGADQEKDKVKMKGYNPAEVGAAGKGYLWKAAGMNMEEEEELQQNLWGLKINMEEESESESEQSMDSEEPDSRGGSPQMDDIKVFQNEVLGTLQRGKEENISCDNLVLEINSLKYAYNISLKEVMQVLSHVVLEFPLQQMDSPLDSSRYCALLLPLLKAWSPVFRNYIKRAADHLEALAAIEDFFLEHEALGISMAKVLMAFYQLEILAEETILSWFSQRDTTDKGQQLRKNQQLQRFIQWLKEAEEESSEDD | |
| 20 μL | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific | |
| 8893 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction