missing translation for 'onlineSavingsMsg'
Learn More

eIF2B epsilon Antibody, DyLight 488, Novus Biologicals™

Product Code. 18424934 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424934 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424934 Supplier Novus Biologicals Supplier No. NBP130448G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

eIF2B epsilon Polyclonal antibody specifically detects eIF2B epsilon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2B epsilon
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CACH, CLE, EIF-2B, eIF-2B GDP-GTP exchange factor subunit epsilon, EIF2BE, EIF2Bepsilon, eukaryotic translation initiation factor 2B, subunit 5 (epsilon, 82kD), eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa, LVWM, translation initiation factor eIF-2B subunit epsilon
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 400 and 450 of human eukaryotic translation initiation factor 2B, subunit 5 epsilon using the numbering given in entry NP_003898.2 (GeneID 8893).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8893
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.