Learn More
Invitrogen™ eIF2b beta Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561636
Description
Immunogen sequence: RETLGLLRQI ITDHRWSNAG ELMELIRREG RRMTAAQPSE TTVGNMVRRV LKIIREEYGR LHGRSDESDQ QESLHKLLTS GGLNEDFSFH YAQLQSNIIE AINELLVELE GTMENIAAQA LEHIHSNEVI MTIGFSRTVE AFLKEAARKR K Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
eIF2b beta is the beta subunit of eukaryotic initiation factor-2B (EIF2B). EIF2B is involved in protein synthesis and exchanges GDP and GTP for its activation and deactivation.
Specifications
| eIF2b beta | |
| Polyclonal | |
| Unconjugated | |
| Eif2b2 | |
| AA409345; C85417; EIF2B; eIF-2B GDP-GTP exchange factor subunit beta; Eif2b2; EIF2BB; EIF-2Bbeta; eukaryotic translation initiation factor 2B subunit beta; eukaryotic translation initiation factor 2B, subunit 2 (beta, 39kD); eukaryotic translation initiation factor 2B, subunit 2 beta; eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; translation initiation factor eIF-2B subunit beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8892 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49770 | |
| Eif2b2 | |
| Recombinant protein corresponding to Human eIF2b beta. Recombinant protein control fragment (Product #RP-103014). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.