missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2b alpha Polyclonal Antibody

eIF2b alpha Polyclonal Antibody
Brand: Invitrogen PA528992
This item is not returnable.
View return policy
Description
eIF2b alpha Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| eIF2b alpha | |
| Polyclonal | |
| Unconjugated | |
| EIF2B1 | |
| 26kDa; D5Ertd406e; eIF-2a; EIF2B; eIF-2B GDP-GTP exchange factor subunit alpha; Eif2b1; EIF2BA; eIF-2Balpha; eukaryotic translation initiation factor 2B subunit alpha; eukaryotic translation initiation factor 2B, subunit 1 (alpha); eukaryotic translation initiation factor 2B, subunit 1 (alpha, 26kDa); eukaryotic translation initiation factor 2B, subunit 1 alpha; eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa; GTP-exchange protein; translation initiation factor eIF-2B subunit alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1967, 209354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14232, Q99LC8 | |
| EIF2B1 | |
| Full length human EIF2B1 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction