Learn More
Invitrogen™ eIF2b alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121151
Description
Positive test controls include: SH-SY5Y, MCF7, SKOV3, NCI-H460, Mouse liver, Mouse brain. Immunogen sequence: MDDKELIEYF KSQMKEDPDM ASAVAAIRTL LEFLKRDKGE TIQGLRANLT SAIETLCGVD SSVAVSSGGE LFLRFISLAS LEYSDYSKCK KIMIERGELF LRRISLSRNK IADLCHTFIK DGATILTHAY SRVVLRVLEA AVAAKKRFSV YVTESQPDLS GKKMAKALCH LNVPVTVVLD AAVGYIMEKA DLVIVGAEGV VENGGIINKI GTNQMAVCAK AQNKPFYVVA ESFKFVRLFP LNQQDVPDKF KYKADTLKVA QTGQDLKEEH PWVDYTAPSL ITLLFTDLGV LTPSAVSDEL IKLYL.
This gene encodes one of five subunits of eukaryotic translation initiation factor 2B (EIF2B), a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
Specifications
| eIF2b alpha | |
| Polyclonal | |
| Unconjugated | |
| EIF2B1 | |
| 26kDa; D5Ertd406e; eIF-2a; EIF2B; eIF-2B GDP-GTP exchange factor subunit alpha; Eif2b1; EIF2BA; eIF-2Balpha; eukaryotic translation initiation factor 2B subunit alpha; eukaryotic translation initiation factor 2B, subunit 1 (alpha); eukaryotic translation initiation factor 2B, subunit 1 (alpha, 26kDa); eukaryotic translation initiation factor 2B, subunit 1 alpha; eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa; GTP-exchange protein; translation initiation factor eIF-2B subunit alpha | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1967, 209354 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.89 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14232, Q99LC8 | |
| EIF2B1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-305 of human EIF2B1 (NP_001405.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.