missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2b alpha Polyclonal Antibody

eIF2b alpha Polyclonal Antibody
Brand: Invitrogen PA598457
This item is not returnable.
View return policy
Description
eIF2b alpha Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| eIF2b alpha | |
| Polyclonal | |
| Unconjugated | |
| EIF2B1 | |
| 26kDa; D5Ertd406e; eIF-2a; EIF2B; eIF-2B GDP-GTP exchange factor subunit alpha; Eif2b1; EIF2BA; eIF-2Balpha; eukaryotic translation initiation factor 2B subunit alpha; eukaryotic translation initiation factor 2B, subunit 1 (alpha); eukaryotic translation initiation factor 2B, subunit 1 (alpha, 26kDa); eukaryotic translation initiation factor 2B, subunit 1 alpha; eukaryotic translation initiation factor 2B, subunit 1 alpha, 26kDa; GTP-exchange protein; translation initiation factor eIF-2B subunit alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1967, 209354 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.78 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14232, Q99LC8 | |
| EIF2B1 | |
| Recombinant Human Translation initiation factor eIF-2B subunit alpha protein (1-305AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction