missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EIF2A Recombinant Rabbit Monoclonal Antibody (23GB1455), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553163
This item is not returnable.
View return policy
Description
EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.
Specifications
| EIF2A | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BJW6, Q9BY44 | |
| EIF2A | |
| A synthesized peptide derived from human eIF2A (450-510AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1455 | |
| Unconjugated | |
| EIF2A | |
| 0910001O23Rik; 2410026C18Rik; 35kDa; 65 kDa eukaryotic translation initiation factor 2A; CDA02; D030048D22; D3Ertd194e; EIF2; EIF-2; EIF2A; eIF-2A; eIF2alpha; eIF-2alpha; eIF-2-alpha; EIF2S1; EIF2S1 protein; eukaryotic translation initiation factor 2 subunit 1; Eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; Eukaryotic translation initiation factor 2A, N-terminally processed; eukaryotic translation initiation factor subunit 1-like protein; initiation factor 2 alpha; MST089; MSTP004; MSTP089 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 229317, 502531, 83939 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction