missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11233-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human EIF2A, recognizes specifically the 65 kDa EIF2A protein.
EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.Specifications
| EIF2A | |
| Polyclonal | |
| Unconjugated | |
| EIF2A | |
| CDA02, EIF 2A, EIF2A, MST089, MSTP004, MSTP089 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229317, 502531, 83939 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BJW6, Q9BY44 | |
| Eif2a | |
| EIF2A Fusion Protein Ag1753 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction