missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2A Mouse anti-Human, Clone: 1G7H4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66482-1-IG-20UL
This item is not returnable.
View return policy
Description
EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.Specifications
| EIF2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BY44 | |
| EIF2A | |
| EIF2A Fusion Protein Ag22041 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 1G7H4 | |
| Unconjugated | |
| EIF2A | |
| CDA02, EIF 2A, EIF2A, MST089, MSTP004, MSTP089 | |
| Mouse | |
| Protein G | |
| RUO | |
| 83939 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction