missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35436-20ul
This item is not returnable.
View return policy
Description
EIF2 beta Polyclonal antibody specifically detects EIF2 beta in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| EIF2 beta | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DKFZp686L18198, EIF2, EIF2B, eIF-2-beta, EIF2beta, eukaryotic initiation factor 2-beta, eukaryotic translation initiation factor 2 beta, eukaryotic translation initiation factor 2 subunit 2, Eukaryotic translation initiation factor 2 subunit beta, eukaryotic translation initiation factor 2, subunit 2 (beta, 38kD ), eukaryotic translation initiation factor 2, subunit 2 beta, 38kDa, MGC8508 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-175 of human EIF2 beta (NP_003899.2).,, Sequence:, MSGDEMIFDPTMSKKKKKKKKPFMLDEEGDTQTEETQPSETKEVEPEPTEDKDLEADEEDTRKKDASDDLDDLNFFNQKKKKKKTKKIFDIDEAEEGVKDLKIESDVQEPTEPEDDLDIMLGNKKKKKKNVKFPDEDEILEKDEALEDEDNKKDDGISFSNQTGPAWAGSERDYT | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8894 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction