missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2 alpha/EIF2S1 Antibody (5E10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97936AF647
This item is not returnable.
View return policy
Description
eIF2 alpha/EIF2S1 Monoclonal specifically detects eIF2 alpha/EIF2S1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| eIF2 alpha/EIF2S1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EIF2S1 | |
| Recombinant human EIF2S1 (1-315aa) purified from E. coli | |
| 100 μL | |
| Phospho Specific | |
| 1965 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 50000000000 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human EIF2S1 mAb, clone 5E10, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human EIF2S1 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction