missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF1AY Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA576163

Product Code. 15992685

  • £472.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

eIF1AY
Polyclonal
Unconjugated
EIF1AY
1500010B24Rik; Ab1-287; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; eIF-1A Y isoform; EIF1AP1; EIF1AX; EIF1AY; EIF4C; eIF-4C; Eukaryotic translation initiation factor 1A; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y chromosome; eukaryotic translation initiation factor 1A, Y-chromosomal; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Liver regeneration-related protein LRRG048; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A
Rabbit
Antigen affinity chromatography
RUO
9086
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O14602
EIF1AY
Recombinant fusion protein of human EIF1AY.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.