missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF1AX Recombinant Rabbit Monoclonal Antibody (23GB5285), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552833
This item is not returnable.
View return policy
Description
This gene encodes an essential eukaryotic translation initiation factor. The protein is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
Specifications
| eIF1AX | |
| Recombinant Monoclonal | |
| Unconjugated | |
| EIF1AX | |
| 1500010B24Rik; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; EIF1AP1; EIF1AX; Eif1ay; EIF4C; eIF-4C; eukaryotic translation initiation factor 1A X-linked; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1964, 302697, 66235 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5285 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P47813, Q8BMJ3 | |
| EIF1AX | |
| A synthesized peptide derived from human eIF1A (60-100AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction