missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF1A Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98066FR
This item is not returnable.
View return policy
Description
EIF1A Polyclonal specifically detects EIF1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EIF1A | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| eIF-1A, EIF1AP1, eIF-4C, EIF4C, eukaryotic translation initiation factor 1A, X chromosome, eukaryotic translation initiation factor 1A, X-chromosomal, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, Eukaryotic translation initiation factor 4C, FLJ45762, Putative eukaryotic translation initiation factor 1A | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 1964 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| EIF1AX | |
| Produced in rabbits immunized with E. coli-derived Human EIF1AY fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction