missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EID2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525543
This item is not returnable.
View return policy
Description
EID2 Polyclonal Antibody for Western Blot
EID2 interacts with EP300 and acts as a repressor of MYOD-dependent transcription and muscle differentiation. Inhibits EP300 histone acetyltransferase activity. Acts as a repressor of TGFB/SMAD transcriptional responses. It may act as a repressor of the TGFB/SMAD3-dependent signaling by selectively blocking formation of TGFB-induced SMAD3-SMAD4 complex.
Specifications
| EID2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6X7S9, Q8N6I1 | |
| EID2 | |
| KLH conjugated synthetic peptide between 177-208 amino acids from the C-terminal region of human EID2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| EID2 | |
| CREBBP/EP300 inhibitor 2; CREBBP/EP300 inhibitory protein 2; Cri2; E1A like inhibitor of differentiation 2; EID-1-like inhibitor of differentiation 2; Eid2; EID-2; EP300 interacting inhibitor of differentiation 2; EP300-interacting inhibitor of differentiation 2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 163126, 386655 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction