missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EID1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97540MFV450
This item is not returnable.
View return policy
Description
EID1 Polyclonal antibody specifically detects EID1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| EID1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| 21 kDa pRb-associated protein, C15orf3PTD014, CREBBP/EP300 inhibitor 1, CREBBP/EP300 inhibitory protein 1MGC138884, CRI1EP300-interacting inhibitor of differentiation 1, E1A-like inhibitor of differentiation 1, EID-1MGC138883, EP300 interacting inhibitor of differentiation 1, IRO45620, NB4 apoptosis related protein, Rb- and p300-binding protein EID-1, RBP21, retinoblastoma protein-associated protein | |
| Produced in rabbits immunized with E. coli-derived Human EID1 fragment. | |
| 0.1 mL | |
| Vision | |
| 23741 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction