missing translation for 'onlineSavingsMsg'
Learn More

Eicosapentaenoic Acid, MedChemExpress

Product Code. 16494437
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494437 50 mg 50mg
16488406 100 mg 100mg
16412196 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494437 Supplier Medchem Express Supplier No. HYB0660
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Eicosapentaenoic Acid (EPA) is an orally active Omega-3 long-chain polyunsaturated fatty acid (ω-3 LC-PUFA).

Eicosapentaenoic Acid exhibits a DNA demethylating action that promotes the re-expression of the tumor suppressor gene CCAAT/enhancer-binding protein δ (C/EBPδ). Eicosapentaenoic Acid activates RAS/ERK/C/EBPβ pathway through H-Ras intron 1 CpG island demethylation in U937 leukemia cells. Eicosapentaenoic Acid can promote relaxation of vascular smooth muscle cells and vasodilation.

Specifications

Chemical Name or Material Eicosapentaenoic Acid
CAS 10417-94-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C20H30O2
Quantity 50 mg
Synonym EPA Timnodonic acid
Solubility Information Ethanol : ≥ 100 mg/mL (330.63 mM) ∣DMSO : ≥ 53 mg/mL (175.24 mM)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O
Molecular Weight (g/mol) 302.45
Formula Weight 302.45
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.