missing translation for 'onlineSavingsMsg'
Learn More

Eicosapentaenoic Acid Ethyl Ester, TRC

Product Code. 30372950
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372950 100 mg 100mg
30383490 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372950 Supplier Toronto Research Chemicals Supplier No. TRCE476880100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eicosanoic acid-ethyl ester
CAS 73310-10-8
Molecular Formula C22 H44 O2
Quantity 100 mg
Synonym EPA 90E, Incromega E 7010
InChI Formula InChI=1S/C22H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24-4-2/h5-6,8-9,11-12,14-15,17-18H,3-4,7,10,13,16,19-21H2,1-2H3/b6-5-,9-8-,12-11-,15-14-,18-17-
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OCC
Molecular Weight (g/mol) 330.5
Formula Weight 330.26
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.