missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eicosapentaenoic acid ethyl ester, MedChemExpress
Description
Eicosapentaenoic acid ethyl ester is an omega-3 fatty acid agent.
Specifications
Specifications
| Chemical Name or Material | Eicosapentaenoic acid ethyl ester |
| Physical Form | Liquid |
| CAS | 86227-47-6 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H34O2 |
| For Use With (Application) | Metabolism-sugar/lipid metabolism |
| Synonym | EPA ethyl ester Ethyl eicosapentaenoate |
| Solubility Information | Ethanol : 50 mg/mL (151.29 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (100.85 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?