missing translation for 'onlineSavingsMsg'
Learn More

Eicosapentaenoic acid ethyl ester, MedChemExpress

Product Code. 16484467
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16484467

Brand: Medchem Express HYB0747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eicosapentaenoic acid ethyl ester is an omega-3 fatty acid agent.

Eicosapentaenoic acid ethyl ester is an omega-3 fatty acid agent.

Specifications

Chemical Name or Material Eicosapentaenoic acid ethyl ester
Physical Form Liquid
CAS 86227-47-6
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H34O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym EPA ethyl ester Ethyl eicosapentaenoate
Solubility Information Ethanol : 50 mg/mL (151.29 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (100.85 mM; Need ultrasonic)
SMILES CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCC(OCC)=O
Molecular Weight (g/mol) 330.5
Formula Weight 330.5
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.