missing translation for 'onlineSavingsMsg'
Learn More

Eicosaacetylmaltohexaose, TRC

Product Code. 30404540
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404540 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404540 Supplier Toronto Research Chemicals Supplier No. TRCE9751651G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eicosaacetylmaltohexaose
CAS 102671-24-9
Molecular Formula C76H102O51
Quantity 1 g
Synonym Tetraacetate-O-2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl-(1-4)-O-2,3,6-tri-O-acetyl-alpha-D-glucopyranosyl-(1-4)-O-2,3,6-tri-O-acetyl-alpha-D-glucopyranosyl-(1-4)-O-2,3,6-tri-O-acetyl-alpha-D-glucopyranosyl-(1-4)-O-2,3,6-tri-O-acetyl-alpha-D-glucopyranosyl-(1-4)-D-glucopyranose
InChI Formula InChI=1S/C76H102O51/c1-27(77)97-21-47-53(103-33(7)83)59(104-34(8)84)66(111-41(15)91)72(118-47)124-55-49(23-99-29(3)79)120-74(68(113-43(17)93)61(55)106-36(10)86)126-57-51(25-101-31(5)81)122-76(70(115-45(19)95)63(57)108-38(12)88)127-58-52(26-102-32(6)82)121-75(69(114-44(18)94)64(58)109-39(13)89)125-56-50(24-100-30(4)80)119-73(67(112-42(16)92)62(56)107-37(11)87)123-54-48(22-98-28(2)78)117-71(116-46(20)96)65(110-40(14)90)60(54)105-35(9)85/h47-76H,21-26H2,1-20H3/t47-,48-,49-,50-,51-,52-,53-,54-,55-,56-,57-,58-,59+,60+,61+,62+,63+,64+,65-,66-,67-,68-,69-,70-,71?,72-,73-,74-,75-,76-/m1/s1
SMILES CC(=O)OC[C@H]1O[C@H](O[C@@H]2[C@@H](COC(=O)C)O[C@H](O[C@@H]3[C@@H](COC(=O)C)OC(OC(=O)C)[C@H](OC(=O)C)[C@H]3OC(=O)C)[C@H](OC(=O)C)[C@H]2OC(=O)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1O[C@H]4O[C@H](COC(=O)C)[C@@H](O[C@H]5O[C@H](COC(=O)C)[C@@H](O[C@H]6O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]6OC(=O)C)[C@H](OC(=O)C)[C@H]5OC(=O)C)[C@H](OC(=O)C)[C@H]4OC(=O)C
IUPAC Name [(2R,3R,4S,5R,6R)-4,5-diacetyloxy-3-[(2R,3R,4S,5R,6R)-3,4-diacetyloxy-6-(acetyloxymethyl)-5-[(2R,3R,4S,5R,6R)-3,4-diacetyloxy-6-(acetyloxymethyl)-5-[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyoxan-2-yl]oxyoxan-2-yl]oxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-[(2R,3R,4S,5R)-4,5,6-triacetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxyoxan-3-yl]oxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol) 1831.59
Formula Weight 1830.5388
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.