missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EI2BL Antibody (OTI9H7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47857
This item is not returnable.
View return policy
Description
EI2BL Monoclonal specifically detects EI2BL in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| EI2BL | |
| Monoclonal | |
| 1.18 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| MRI1 | |
| Full length human recombinant protein of human MRI1 (NP_115661) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 84245 | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI9H7 | |
| Unconjugated | |
| EC 5.3.1.23, M1Pi, Mediator of RhoA-dependent invasion, methylthioribose-1-phosphate isomerase, methylthioribose-1-phosphate isomerase homolog (S. cerevisiae), MGC3207, MRDI, MTNA, MTR-1-P isomerase, S-methyl-5-thioribose-1-phosphate isomerase, Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein, Ypr118w | |
| Mouse | |
| 39.1 kDa | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for Homo sapiens methylthioribose-1-phosphate isomerase homolog (S. cerevisiae) (MRI1), transcript variant 1. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction