missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EI2BL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35428-100ul
This item is not returnable.
View return policy
Description
EI2BL Polyclonal antibody specifically detects EI2BL in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| EI2BL | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 5.3.1.23, M1Pi, Mediator of RhoA-dependent invasion, methylthioribose-1-phosphate isomerase, methylthioribose-1-phosphate isomerase homolog (S. cerevisiae), MGC3207, MRDI, MTNA, MTR-1-P isomerase, S-methyl-5-thioribose-1-phosphate isomerase, Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein, Ypr118w | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-369 of human EI2BL (NP_001026897.1).,, Sequence:, MTLEAIRYSRGSLQILDQLLLPKQSRYEAVGSVHQAWEAIRAMKVRGAPAIALVGCLSLAVELQAGAGGPGLAALVAFVRDKLSFLVTARPTAVNMARAARDLADVAAREAEREGATEEAVRERVICCTEDMLEKDLRDNRSIGDLGARHLLERVAPSGGKVTVLTHCNTGALATAGYGTALGVIRSLHSLGRLEHAFCTETRPYNQGARLTAFELVYEQIPATLITDSMVAAAMAHRGVSAVVVGADRVVANGDTANKVGTYQLAIVAKHHGIPFYVAAPSSSCDLRLETGKEIIIEERPGQELTDVNGVRIAAPGIGVWNPAFDVTPHDLITGGIITELGVFAPEELRTALTTTISSRDGTLDGPQM | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84245 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction