missing translation for 'onlineSavingsMsg'
Learn More

EI2BL Antibody, DyLight 594, Novus Biologicals™

Product Code. 18349946 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349946 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349946 Supplier Novus Biologicals Supplier No. NBP306310DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EI2BL Polyclonal specifically detects EI2BL in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EI2BL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 5.3.1.23, M1Pi, Mediator of RhoA-dependent invasion, methylthioribose-1-phosphate isomerase, methylthioribose-1-phosphate isomerase homolog (S. cerevisiae), MGC3207, MRDI, MTNA, MTR-1-P isomerase, S-methyl-5-thioribose-1-phosphate isomerase, Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein, Ypr118w
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EI2BL fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84245
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.