missing translation for 'onlineSavingsMsg'
Learn More

EI24 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18419475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419475 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419475 Supplier Novus Biologicals Supplier No. NBP281820AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EI24 Polyclonal antibody specifically detects EI24 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen EI24
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias EPG4, etoposide induced 2.4 mRNA, p53-induced gene 8 protein, PIG8etoposide-induced protein 2.4 homolog, TP53I8, tumor protein p53 inducible protein 8
Host Species Rabbit
Immunogen EI24 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human EI24. The immunogen is located within the last 50 amino acids of EI24. Amino Acid Squence: STSAEKFPSPHPSPAKLKA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9538
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.