missing translation for 'onlineSavingsMsg'
Learn More

EI1, MedChemExpress

Product Code. 16499197
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499197

Brand: Medchem Express HY15573

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EI1 (KB-145943) is a potent and selective EZH2 inhibitor with IC50 of 15 nM and 13 nM for EZH2 (WT) and EZH2 (Y641F), respectively.

EI1 (KB-145943) is a potent and selective EZH2 inhibitor with IC50 of 15 nM and 13 nM for EZH2 (WT) and EZH2 (Y641F), respectively.

Specifications

Chemical Name or Material EI1
CAS 1418308-27-6
Color White
Purity Grade Notes Research
Molecular Formula C23H26N4O2
Quantity 5 mg
Synonym KB-145943
Solubility Information DMSO : 7.69 mg/mL (19.69 mM; Need ultrasonic)
SMILES CCC(CC)N(C=C1)C2=C1C(C(NCC3=C(C)C=C(C)NC3=O)=O)=CC(C#N)=C2
Molecular Weight (g/mol) 390.48
Formula Weight 390.48
Percent Purity 99.18%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.