missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EHop 016
GREENER_CHOICE

Product Code. p-7116115
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18738346

Brand: Tocris Bioscience™ 6248/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rac inhibitor

EHop 016 is a Rac inhibitor (IC50 = 1.1 μM). Exhibits some selectivity for Rac1 and Rac3 over Cdc42. Blocks the interaction of Rac with guanine nucleotide exchange factor Vav2 in breast cancer cell lines expressing high levels of Rac activity and inhibits activity of Rac downstream effector PAK1. Also inhibits mammary tumor growth and metastasis in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rho GTPase Inhibitors
CAS 1380432-32-5
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C25H30N6O
Formula Weight Observed MW: 430.55
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.