missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EHop-016, MedChemExpress
Description
EHop-016 is a potent and selective Rac GTPase Rac1 and Rac3 inhibitor. EHop-016 inhibits Rac1 activity with an IC50 of 1.1 μM in MDA-MB-435 cells. EHop-016 inhibits Vav2 interaction with Rac, Rac-activated PAK1, lamellipodia formation, and cell migration.
Specifications
Specifications
| Chemical Name or Material | EHop-016 |
| CAS | 1380432-32-5 |
| Color | Pink |
| Purity Grade Notes | Research |
| Molecular Formula | C25H30N6O |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 32 mg/mL (74.32 mM) |
| SMILES | CCN1C2=C(C3=C1C=CC=C3)C=C(NC4=NC(NCCCN5CCOCC5)=NC=C4)C=C2 |
| Molecular Weight (g/mol) | 430.55 |
| Formula Weight | 430.55 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?