missing translation for 'onlineSavingsMsg'
Learn More

EHop-016, MedChemExpress

Product Code. 16445368
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16445368

Brand: Medchem Express HY12810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EHop-016 is a potent and selective Rac GTPase Rac1 and Rac3 inhibitor. EHop-016 inhibits Rac1 activity with an IC50 of 1.1 μM in MDA-MB-435 cells. EHop-016 inhibits Vav2 interaction with Rac, Rac-activated PAK1, lamellipodia formation, and cell migration.

EHop-016 is a potent and selective Rac GTPase Rac1 and Rac3 inhibitor. EHop-016 inhibits Rac1 activity with an IC50 of 1.1 μM in MDA-MB-435 cells. EHop-016 inhibits Vav2 interaction with Rac, Rac-activated PAK1, lamellipodia formation, and cell migration.

Specifications

Chemical Name or Material EHop-016
CAS 1380432-32-5
Color Pink
Purity Grade Notes Research
Molecular Formula C25H30N6O
Quantity 10 mg
Solubility Information DMSO : ≥ 32 mg/mL (74.32 mM)
SMILES CCN1C2=C(C3=C1C=CC=C3)C=C(NC4=NC(NCCCN5CCOCC5)=NC=C4)C=C2
Molecular Weight (g/mol) 430.55
Formula Weight 430.55
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.