missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EHop 016
GREENER_CHOICE

Product Code. 18738346
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738346 25 mg 25mg
18709786 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18738346 Supplier Tocris Bioscience™ Supplier No. 6248/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rac inhibitor

EHop 016 is a Rac inhibitor (IC50 = 1.1 μM). Exhibits some selectivity for Rac1 and Rac3 over Cdc42. Blocks the interaction of Rac with guanine nucleotide exchange factor Vav2 in breast cancer cell lines expressing high levels of Rac activity and inhibits activity of Rac downstream effector PAK1. Also inhibits mammary tumor growth and metastasis in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rho GTPase Inhibitors
CAS 1380432-32-5
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C25H30N6O
Formula Weight Observed MW: 430.55
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.