missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EHNA hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective inhibitor of cGMP-stimulated phosphodiesterase (PDE2) (IC50 = 0.8 - 4 μM). Also a potent inhibitor of adenosine deaminase. Suppresses spontaneous differentiation of human ESCs in feeder-free conditions. Also prevents directed neuronal differentiation.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 313.83 |
| Purity | >98% |
| Molecular Weight (g/mol) | 313.83 |
| Chemical Name or Material | EHNA hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction