missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EHMT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10008799
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10008799 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10008799 Supplier Invitrogen™ Supplier No. PA511208

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EHMT1 Polyclonal Antibody for Western Blot

EHMT1, also known as EUHMTASE1, is a histone methyltransferase. This protein methylates 'Lys-9' of histone H3 in vitro. H3 'Lys-9' methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. EHMT1 is Probably targeted to histone H3 by different DNA-binding proteins like E2F6, MGA, MAX and/or DP1. During G0 phase, it probably contributes to silencing of MYC- and E2F-responsive genes, suggesting a role in the G0/G1 transition of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHMT1
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EHMT1
Gene Accession No. Q9H9B1
Gene Alias 9230102N17Rik; bA188C12.1; D330003E03; EHMT1; euchromatic histone lysine methyltransferase 1; euchromatic histone methyltransferase 1; euchromatic histone-lysine N-methyltransferase 1; Euhmtase1; Eu-HMTase1; FP13812; G9a-like protein 1; GLP; GLP1; H3-K9-HMTase 5; histone H3-K9 methyltransferase 5; Histone-lysine N-methyltransferase EHMT1; histone-lysine N-methyltransferase, H3 lysine-9 specific 5; KIAA1876; KMT1D; lysine N-methyltransferase 1D; mKIAA1876
Gene Symbols EHMT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 195-226 amino acids from the N-terminal region of human EHMT1 (EUHMTASE1).
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79813
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.