missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511134
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
EHMT1, also known as EUHMTASE1, is a histone methyltransferase. This protein methylates 'Lys-9' of histone H3 in vitro. H3 'Lys-9' methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. EHMT1 is Probably targeted to histone H3 by different DNA-binding proteins like E2F6, MGA, MAX and/or DP1. During G0 phase, it probably contributes to silencing of MYC- and E2F-responsive genes, suggesting a role in the G0/G1 transition of the cell cycle.
Specifications
| EHMT1 | |
| Polyclonal | |
| Unconjugated | |
| EHMT1 | |
| 9230102N17Rik; bA188C12.1; D330003E03; EHMT1; euchromatic histone lysine methyltransferase 1; euchromatic histone methyltransferase 1; euchromatic histone-lysine N-methyltransferase 1; Euhmtase1; Eu-HMTase1; FP13812; G9a-like protein 1; GLP; GLP1; H3-K9-HMTase 5; histone H3-K9 methyltransferase 5; Histone-lysine N-methyltransferase EHMT1; histone-lysine N-methyltransferase, H3 lysine-9 specific 5; KIAA1876; KMT1D; lysine N-methyltransferase 1D; mKIAA1876 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 79813 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H9B1 | |
| EHMT1 | |
| KLH conjugated synthetic peptide between 1145-1174 amino acids from the C-terminal region of human EHMT1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction