missing translation for 'onlineSavingsMsg'
Learn More

EHHADH Antibody (OTI2C4), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18411483 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411483 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411483 Supplier Novus Biologicals Supplier No. NBP270625AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EHHADH Monoclonal antibody specifically detects EHHADH in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHHADH
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C4
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase, enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase, LBFP, LBP, L-PBE, PBFEMGC120586, peroxisomal, peroxisomal bifunctional enzyme3,2-trans-enoyl-CoA isomerase, peroxisomal enoyl-CoA hydratase
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 496-723 of human EHHADH (NP_001957) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1962
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.