missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EHF Recombinant Rabbit Monoclonal Antibody (7O0N9)

Product Code. 30284548
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30284548 20 μL 20µL
30284274 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30284548 Supplier Invitrogen™ Supplier No. MA558133

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MILEGGGVMN LNPGNNLLHQ PPAWTDSYST CNVSSGFFGG QWHEIHPQYW TKYQVWEWLQ HLLDTNQLDA NCIPFQEFDI NGEHLCSMSL QEFTRAAGTA GQLLYSNLQH LKW.

This gene encodes a protein that belongs to an ETS transcription factor subfamily characterized by epithelial-specific expression (ESEs). The encoded protein acts as a transcriptional repressor and may be associated with asthma susceptibility. This protein may be involved in epithelial differentiation and carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EHF
Applications ELISA, Immunoprecipitation
Classification Recombinant Monoclonal
Clone 7O0N9
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene EHF
Gene Accession No. Q9NZC4
Gene Alias 9030625L19Rik; AU019492; EHF; Epithelium-specific Ets transcription factor 3; ESE3; ESE-3; ESE3 transcription factor; ESE3B; ESEJ; ETS domain-containing transcription factor; ETS homologous factor; hEHF; Unknown (protein for MGC:133971)
Gene Symbols EHF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-113 of human EHF (NP_036285.2).
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26298
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.